Tisk…

3-Butynyl toluene-4-sulphonate

Dodavatel: Apollo Scientific

Synonyms: 3-Butynyl 4-methylbenzenesulphonate, 3-Butynyl p-toluenesulphonate

OR7595-25G OR7595-100G
APOSOR7595-25GEA 5100 CZK
APOSOR7595-25G APOSOR7595-100G
3-Butynyl toluene-4-sulphonate
3-Butynyl toluene-4-sulphonate
Vzorec: C₁₁H₁₂O₃S
MW: 224.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041687
CAS číslo: 23418-85-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR