Tisk…

3-Bromo-5-chloroaniline 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR7300-10G OR7300-5G OR7300-25G
APOSOR7300-10GEA 1290 CZK
APOSOR7300-10G APOSOR7300-5G APOSOR7300-25G
3-Bromo-5-chloroaniline 98%
3-Bromo-5-chloroaniline
Vzorec: C₆H₅BrClN
Storage Temperature: Refrigerator
MDL Number: MFCD06797804
CAS číslo: 96558-78-0
UN: 2811
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR