Tisk…

3-Vinylphenylboronic acid 95%

Dodavatel: Apollo Scientific

Synonyms: Dihydroxy(3-vinylphenyl)borane, 3-Vinylbenzeneboronic acid, (3-Vinylphenyl)boronic acid, (3-Vinylphenyl)metaboric acid

OR7268-5G
APOSOR7268-5GEA 3610 CZK
APOSOR7268-5G
3-Vinylphenylboronic acid 95%
3-Vinylphenylboronic acid
Vzorec: C₈H₉BO₂
MW: 147.97
Bod tání: 141…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074679
CAS číslo: 15016-43-0
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR