Tisk…

Diisopropyl(bromomethyl)boronate 95%

Dodavatel: Apollo Scientific
Varování

OR7201-1G OR7201-5G
APOSOR7201-1GEA 5390 CZK
APOSOR7201-1G APOSOR7201-5G
Diisopropyl(bromomethyl)boronate 95%
Diisopropyl(bromomethyl)boronate
Vzorec: C₇H₁₆BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD01631222
CAS číslo: 137297-49-5
UN: 3272
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR