Tisk…

5-Chloro-2-iodophenol

Dodavatel: Apollo Scientific
Varování

OR70040-1G OR70040-5G
APOSOR70040-1GEA 1490 CZK
APOSOR70040-1G APOSOR70040-5G
5-Chloro-2-iodophenol
5-Chloro-2-iodophenol
Vzorec: C₆H₄ClIO
Storage Temperature: Refrigerator
MDL Number: MFCD09753473
CAS číslo: 136808-72-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR