Tisk…

(1S)-(-)-Camphanic chloride

Dodavatel: Apollo Scientific

OR6501T-10G OR6501T-5G
APOSOR6501T-10GEA 9330 CZK
APOSOR6501T-10G APOSOR6501T-5G
(1S)-(-)-Camphanic chloride
(1S)-(-)-Camphanic chloride

An optically active resolving agent.

Vzorec: C₁₀H₁₃ClO₃
Bod tání: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00135626
CAS číslo: 39637-74-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR