Tisk…

3-Formyl-2-methyl-5-nitroindole 95+%

Dodavatel: Apollo Scientific
Varování

OR6332-1G OR6332-5G OR6332-500MG
APOSOR6332-1GEA 2280 CZK
APOSOR6332-1G APOSOR6332-5G APOSOR6332-500MG
3-Formyl-2-methyl-5-nitroindole 95+%
3-Formyl-2-methyl-5-nitroindole
Vzorec: C₁₀H₈N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00216481
CAS číslo: 3558-17-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR