Tisk…

tert-Butyl-4-formylpyridin-3-ylcarbamate 97%

Dodavatel: Apollo Scientific
Varování

OR6174-10G OR6174-1G OR6174-5G
APOSOR6174-10GEA 32670 CZK
APOSOR6174-10G APOSOR6174-1G APOSOR6174-5G
tert-Butyl-4-formylpyridin-3-ylcarbamate 97%
tert-Butyl-4-formylpyridin-3-ylcarbamate
Vzorec: C₁₁H₁₄N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD02683209
CAS číslo: 116026-95-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR