Tisk…

5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2-methyl-1H-imidazo[4,5-b]pyridine, 6-Bromo-2-methyl-7-azabenzimidazole

OR61523-1G OR61523-250MG
APOSOR61523-1GEA 5630 CZK
APOSOR61523-1G APOSOR61523-250MG
5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
Vzorec: C₇H₆BrN₃
MW: 212.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16657201
CAS číslo: 219762-28-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR