Tisk…

N-(Toluene-4-sulphonyl)glycine methyl ester

Dodavatel: Apollo Scientific

OR61063-100G OR61063-25G OR61063-500G
APOSOR61063-100GEA 3270 CZK
APOSOR61063-100G APOSOR61063-25G APOSOR61063-500G
N-(Toluene-4-sulphonyl)glycine methyl ester
N-(Toluene-4-sulphonyl)glycine methyl ester
Vzorec: C₁₀H₁₃NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00558890
CAS číslo: 2645-02-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR