Tisk…

4-Bromo-2-chloro-6-nitroaniline

Dodavatel: Apollo Scientific
Varování

OR60213-1G OR60213-250MG OR60213-5G
APOSOR60213-1GEA 910 CZK
APOSOR60213-1G APOSOR60213-250MG APOSOR60213-5G
4-Bromo-2-chloro-6-nitroaniline
4-Bromo-2-chloro-6-nitroaniline
Vzorec: C₆H₄BrClN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD03426655
CAS číslo: 34033-41-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR