Tisk…

2-Amino-4-chlorothiazole-5-carbaldehyde 98%

Dodavatel: Apollo Scientific
Varování

OR59894-1G OR59894-5G OR59894-10G
APOSOR59894-1GEA 953 CZK
APOSOR59894-1G APOSOR59894-5G APOSOR59894-10G
2-Amino-4-chlorothiazole-5-carbaldehyde 98%
2-Amino-4-chlorothiazole-5-carbaldehyde
Vzorec: C₄H₃ClN₂OS
Storage Temperature: Refrigerator
MDL Number: MFCD00204993
CAS číslo: 76874-79-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR