Tisk…

Methyl (4-bromo-1H-pyrazol-1-yl)acetate

Dodavatel: Apollo Scientific

OR59391-5G
APOSOR59391-5GEA 3940 CZK
APOSOR59391-5G
Methyl (4-bromo-1H-pyrazol-1-yl)acetate
Methyl (4-bromo-1H-pyrazol-1-yl)acetate
Vzorec: C₆H₇BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09878400
CAS číslo: 1072944-71-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR