Tisk…

1-Boc-7-methylindole

Dodavatel: Apollo Scientific

OR59346-1G OR59346-5G
APOSOR59346-1GEA 5330 CZK
APOSOR59346-1G APOSOR59346-5G
1-Boc-7-methylindole
1-Boc-7-methylindole
Vzorec: C₁₄H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD09878411
CAS číslo: 442910-62-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR