Tisk…

3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid 97%

Dodavatel: Apollo Scientific

OR5816-1G
APOSOR5816-1GEA 1990 CZK
APOSOR5816-1G
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid 97%
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid
Vzorec: C₁₀H₁₁BClNO₃
Bod tání: 216…220 °C
Storage Temperature: Ambient
MDL Number: MFCD07363770
CAS číslo: 850589-44-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR