Tisk…

2'-(Methylsulphonyl)acetanilide

Dodavatel: Apollo Scientific
Varování

OR5729-1G OR5729-25G OR5729-5G
APOSOR5729-1GEA 622 CZK
APOSOR5729-1G APOSOR5729-25G APOSOR5729-5G
2'-(Methylsulphonyl)acetanilide
2'-(Methylsulphonyl)acetanilide
Vzorec: C₉H₁₁NO₃S
Storage Temperature: Ambient
MDL Number: MFCD01464107
CAS číslo: 20628-27-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR