Tisk…

8-hydroxyquinoline-2-carbonitrile 99%

Dodavatel: Apollo Scientific
Varování

OR55719-1G OR55719-250MG OR55719-5G
APOSOR55719-1GEA 3110 CZK
APOSOR55719-1G APOSOR55719-250MG APOSOR55719-5G
8-hydroxyquinoline-2-carbonitrile 99%
8-hydroxyquinoline-2-carbonitrile
Vzorec: C₁₀H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD00216717
CAS číslo: 6759-78-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR