Tisk…

2-(tert-Butoxy)-2-oxoethylzinc bromide 0.5M in diethyl ether

Dodavatel: Apollo Scientific
Nebezpečí

OR55579-100ML OR55579-50ML
APOSOR55579-100MLEA 41660 CZK
APOSOR55579-100ML APOSOR55579-50ML
2-(tert-Butoxy)-2-oxoethylzinc bromide 0.5M in diethyl ether
2-(tert-Butoxy)-2-oxoethylzinc bromide
Vzorec: C₆H₁₁BrO₂Zn
Bod vzplanutí: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671971
CAS číslo: 51656-70-3
UN: 1993
ADR: 3,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR