Tisk…

6-Nitroanthranilic acid

Dodavatel: Apollo Scientific
Varování

OR55396-1G OR55396-25G OR55396-5G
APOSOR55396-1GEA 622 CZK
APOSOR55396-1G APOSOR55396-25G APOSOR55396-5G
6-Nitroanthranilic acid
6-Nitroanthranilic acid
Vzorec: C₇H₆N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00034785
CAS číslo: 50573-74-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR