Tisk…

4-Hydroxy-3-isopropylphenylboronic acid, pinacol ester

Dodavatel: Apollo Scientific
Varování

OR55387-500MG
APOSOR55387-500MGEA 12440 CZK
APOSOR55387-500MG
4-Hydroxy-3-isopropylphenylboronic acid, pinacol ester
4-Hydroxy-3-isopropylphenylboronic acid, pinacol ester
Vzorec: C₁₅H₂₃BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16994212
CAS číslo: 1188335-75-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR