Tisk…

3-Chloro-(4-ethoxycarbonyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific

Synonyms: (3-Chloro-4-(ethoxycarbonyl)phenyl)boronic acid, 3-Chloro-4-(ethoxycarbonyl)phenylboronic acid

OR5532-1G OR5532-5G
APOSOR5532-1GEA 2780 CZK
APOSOR5532-1G APOSOR5532-5G
3-Chloro-(4-ethoxycarbonyl)phenylboronic acid 98%
3-Chloro-(4-ethoxycarbonyl)phenylboronic acid
Vzorec: C₉H₁₀BClO₄
MW: 228.4373 g/mol
Bod tání: 122…126 °C
Storage Temperature: Ambient
MDL Number: MFCD06659875
CAS číslo: 850568-11-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR