Tisk…

Diethyl 4-iodopyridine-2,6-dicarboxylate 98+%

Dodavatel: Apollo Scientific
Varování

OR55111-1G OR55111-250MG
APOSOR55111-1GEA 10880 CZK
APOSOR55111-1G APOSOR55111-250MG
Diethyl 4-iodopyridine-2,6-dicarboxylate 98+%
Diethyl 4-iodopyridine-2,6-dicarboxylate
Vzorec: C₁₁H₁₂INO₄
Storage Temperature: Ambient
MDL Number: MFCD11042539
CAS číslo: 120491-90-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR