Tisk…

Octadecanal 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Octadecyl Aldehyde, Octadecanaldehyde, Stearyl Aldehyde, Stearaldehyde

OR54467-1G OR54467-5G
APOSOR54467-1GEA 1580 CZK
APOSOR54467-1G APOSOR54467-5G
Octadecanal 95%
Octadecanal
Vzorec: C₁₈H₃₆O
MW: 268.49 g/mol
Bod varu: 215 °C/20mmHg
Bod tání: 46 °C
Storage Temperature: Freezer
MDL Number: MFCD03790007
CAS číslo: 638-66-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR