Tisk…

9-Aminofluorene hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 9-Fluorenamine Hydrochloride, 9H-Fluoren-9-amine hydrochloride

OR54458-1G OR54458-25G OR54458-5G
APOSOR54458-1GEA 1040 CZK
APOSOR54458-1G APOSOR54458-25G APOSOR54458-5G
9-Aminofluorene hydrochloride
9-Aminofluorene hydrochloride
Vzorec: C₁₃H₁₁N
MW: 217.70 g/mol
Bod tání: 265…370 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012536
CAS číslo: 5978-75-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR