Tisk…

2-Bromo-3-nitrobenzonitrile

Dodavatel: Apollo Scientific
Varování

OR54427-1G OR54427-250MG
APOSOR54427-1GEA 17360 CZK
APOSOR54427-1G APOSOR54427-250MG
2-Bromo-3-nitrobenzonitrile
2-Bromo-3-nitrobenzonitrile
Vzorec: C₇H₃BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18392274
CAS číslo: 90407-28-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR