Tisk…

2-Bromobenzenesulphonyl chloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromobenzene-1-sulphonyl chloride

OR5370-100G
APOSOR5370-100GEA 4770 CZK
APOSOR5370-100G
2-Bromobenzenesulphonyl chloride 95%
2-Bromobenzenesulphonyl chloride
Vzorec: C₆H₄BrClO₂S
MW: 255.52
Bod varu: 117 °C/2.25mmHg
Bod tání: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00051971
CAS číslo: 2905-25-1
UN: 3261
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR