Tisk…

Cyclopropanecarboxylic acid 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR5295-1KG OR5295-250G OR5295-50G
APOSOR5295-1KGEA 4070 CZK
APOSOR5295-1KG APOSOR5295-250G APOSOR5295-50G
Cyclopropanecarboxylic acid 98%
Cyclopropanecarboxylic acid
Vzorec: C₄H₆O₂
MW: 86.09
Bod varu: 182…184 °C/760mmHg
Bod tání: 14…19 °C
Hustota: 1.087
Bod vzplanutí: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00001287
CAS číslo: 1759-53-1
EINECS: 217-162-5
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR