Tisk…

1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine 90%

Dodavatel: Apollo Scientific
Varování

OR52575-250MG OR52575-1G OR52575-500MG
APOSOR52575-250MGEA 11480 CZK
APOSOR52575-250MG APOSOR52575-1G APOSOR52575-500MG
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine 90%
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine
Vzorec: C₁₂H₂₂BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16039627
CAS číslo: 1254982-25-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR