Tisk…

(1R)-(-)-Camphor-10-sulphonic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: L(-)-Camphor-10-sulphonic acid, (1R)-(-)-2-Oxobornane-10-sulphonic acid, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (-)-CSA

OR52399-250G
APOSOR52399-250GEA 3940 CZK
APOSOR52399-250G
(1R)-(-)-Camphor-10-sulphonic acid 97%
(1R)-(-)-Camphor-10-sulphonic acid
Vzorec: C₁₀H₁₆O₄S
MW: 232.3
Bod varu: 375
Bod tání: 198 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00150753
CAS číslo: 35963-20-3
EINECS: 252-817-9
UN: 3261
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR