Tisk…

4-(Bromomethyl)benzyl alcohol 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR52377-10G OR52377-1G OR52377-5G
APOSOR52377-10GEA 10490 CZK
APOSOR52377-10G APOSOR52377-1G APOSOR52377-5G
4-(Bromomethyl)benzyl alcohol 97%
4-(Bromomethyl)benzyl alcohol
Vzorec: C₈H₉BrO
Bod tání: 76…77 °C
Storage Temperature: Freezer
MDL Number: MFCD08275218
CAS číslo: 71831-21-5
UN: 3261
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR