Tisk…

(S)-1-(Isoquinolin-6-yl)ethanamine

Dodavatel: Apollo Scientific

OR52295-500MG
APOSOR52295-500MGEA 13260 CZK
APOSOR52295-500MG
(S)-1-(Isoquinolin-6-yl)ethanamine
(S)-1-(Isoquinolin-6-yl)ethanamine
Vzorec: C₁₁H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD21348045
CAS číslo: 1344588-21-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR