Tisk…

Glyoxylic acid monohydrate 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Formylformic acid monohydrate, Oxoacetic acid monohydrate, 2-Oxoethanoic acid monohydrate

OR52099-100G OR52099-1KG OR52099-500G
APOSOR52099-100GEA 1740 CZK
APOSOR52099-100G APOSOR52099-1KG APOSOR52099-500G
Glyoxylic acid monohydrate 97%
Glyoxylic acid monohydrate
Vzorec: C₂H₂O₃·1H₂O
Bod varu: 100 °C
Bod tání: 49…52 °C
Bod vzplanutí: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00127974
CAS číslo: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR