Tisk…

tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate 95%

Dodavatel: Apollo Scientific
Varování

OR51932-25G OR51932-5G
APOSOR51932-25GEA 0 CZK
APOSOR51932-25G APOSOR51932-5G
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate 95%
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate
Vzorec: C₁₁H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08726023
CAS číslo: 146504-07-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR