Tisk…

(-)-Dibenzoyl-L-tartaric acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (2R,3R)-2,3-Bis(benzoyloxy)succinic acid

OR51878-100G
APOSOR51878-100GEA 2440 CZK
APOSOR51878-100G
(-)-Dibenzoyl-L-tartaric acid
(-)-Dibenzoyl-L-tartaric acid
Vzorec: C₁₈H₁₄O₈
MW: 358.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003074
CAS číslo: 2743-38-6
EINECS: 241-097-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR