Tisk…

Fmoc-val-OSu 94%

Dodavatel: Apollo Scientific

Synonyms: N-Fmoc-L-valine N-succinimidyl ester, Fmoc-Val-OSu

OR51362-25G OR51362-100G
APOSOR51362-25GEA 1080 CZK
APOSOR51362-25G APOSOR51362-100G
Fmoc-val-OSu 94%
Fmoc-val-OSu
Vzorec: C₂₄H₂₄N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00153370
CAS číslo: 130878-68-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR