Tisk…

(R,R)-iPr-BI-DIME ≥98%

Dodavatel: Apollo Scientific

Synonyms: (2R,3R)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-isopropyl-2,3-dihydrobenzo[d][1,3]oxaphosphole

OR510256-100MG
APOSOR510256-100MGEA 4470 CZK
APOSOR510256-100MG
(R,R)-iPr-BI-DIME ≥98%
(R,R)-iPr-BI-DIME
Vzorec: C₂₂H₂₉O₃P
MW: 372.438 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31630762
CAS číslo: 1477517-19-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR