Tisk…

Methyl 5-amino-2-methoxy-4-methylbenzoate ≥98%

Dodavatel: Apollo Scientific

OR510246-100MG
APOSOR510246-100MGEA 8900 CZK
APOSOR510246-100MG
Methyl 5-amino-2-methoxy-4-methylbenzoate ≥98%
Methyl 5-amino-2-methoxy-4-methylbenzoate
Vzorec: C₁₀H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD19443224
CAS číslo: 70752-21-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR