Tisk…

7-Chloro-6-nitroquinoline ≥98%

Dodavatel: Apollo Scientific

OR510173-100MG
APOSOR510173-100MGEA 11540 CZK
APOSOR510173-100MG
7-Chloro-6-nitroquinoline ≥98%
7-Chloro-6-nitroquinoline
Vzorec: C₉H₅ClN₂O₂
MW: 208,601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18448459
CAS číslo: 58416-31-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR