Tisk…

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran 99%

Dodavatel: Apollo Scientific

OR51008-1G OR51008-25G OR51008-5G
APOSOR51008-1GEA 744 CZK
APOSOR51008-1G APOSOR51008-25G APOSOR51008-5G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran 99%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran
Vzorec: C₁₈H₁₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD18910632
CAS číslo: 912824-85-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR