Tisk…

Methyl 4-bromo-3-(hydroxymethyl)benzoate ≥95%

Dodavatel: Apollo Scientific

OR510049-250MG
APOSOR510049-250MGEA 3810 CZK
APOSOR510049-250MG
Methyl 4-bromo-3-(hydroxymethyl)benzoate ≥95%
Methyl 4-bromo-3-(hydroxymethyl)benzoate
Vzorec: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527255
CAS číslo: 254746-40-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR