Tisk…

5-Bromoisoquinoline-1-carbonitrile ≥98%

Dodavatel: Apollo Scientific

OR510044-250MG
APOSOR510044-250MGEA 8900 CZK
APOSOR510044-250MG
5-Bromoisoquinoline-1-carbonitrile ≥98%
5-Bromoisoquinoline-1-carbonitrile
Vzorec: C₁₀H₅BrN₂
MW: 233,064 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17215757
CAS číslo: 956003-76-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR