Tisk…

4-Bromo-2,3-dihydrobenzo[b]thiophen-3-ol ≥97%

Dodavatel: Apollo Scientific

OR510041-250MG
APOSOR510041-250MGEA 2110 CZK
APOSOR510041-250MG
4-Bromo-2,3-dihydrobenzo[b]thiophen-3-ol ≥97%
4-Bromo-2,3-dihydrobenzo[b]thiophen-3-ol
Vzorec: C₈H₇BrOS
Storage Temperature: Refrigerator
MDL Number: MFCD27928115
CAS číslo: 737802-12-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR