Tisk…

3-Aminopropan-1-ol 99%

Dodavatel: Apollo Scientific

OR50998-100G OR50998-500G
APOSOR50998-100GEA 1670 CZK
APOSOR50998-100G APOSOR50998-500G
3-Aminopropan-1-ol 99%
3-Aminopropan-1-ol
Vzorec: C₃H₉NO
Storage Temperature: Ambient
MDL Number: MFCD00008223
CAS číslo: 156-87-6
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR