Tisk…

2-Ethoxyacetyl chloride 97%

Dodavatel: Apollo Scientific

OR50944-100G
APOSOR50944-100GEA 4860 CZK
APOSOR50944-100G
2-Ethoxyacetyl chloride 97%
2-Ethoxyacetyl chloride
Vzorec: C₄H₇ClO₂
MW: 122.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11170566
CAS číslo: 14077-58-8
UN: 2920
ADR: 8+3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR