Tisk…

1-tert-Butoxyvinyloxy-tert-butyldimethylsilane 90%

Dodavatel: Apollo Scientific

OR480884-1G OR480884-5G
APOSOR480884-1GEA 5130 CZK
APOSOR480884-1G APOSOR480884-5G
1-tert-Butoxyvinyloxy-tert-butyldimethylsilane 90%
1-tert-Butoxyvinyloxy-tert-butyldimethylsilane
Vzorec: C₁₂H₂₆O₂Si
Storage Temperature: Ambient
MDL Number: MFCD12923224
CAS číslo: 74786-02-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR