Tisk…

tert-Butyl N-[(5-chloro-2-pyridyl)methyl]carbamate

Dodavatel: Apollo Scientific

OR480812-1G OR480812-5G
APOSOR480812-1GEA 4860 CZK
APOSOR480812-1G APOSOR480812-5G
tert-Butyl N-[(5-chloro-2-pyridyl)methyl]carbamate
tert-Butyl N-[(5-chloro-2-pyridyl)methyl]carbamate
Vzorec: C₁₁H₁₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD19982297
CAS číslo: 67938-77-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR