Tisk…

5-Bromo-2-cyclopropylaniline

Dodavatel: Apollo Scientific

OR48081-1G OR48081-250MG OR48081-5G
APOSOR48081-1GEA 16370 CZK
APOSOR48081-1G APOSOR48081-250MG APOSOR48081-5G
5-Bromo-2-cyclopropylaniline
5-Bromo-2-cyclopropylaniline
Vzorec: C₉H₁₀BrN
Storage Temperature: Freezer
MDL Number: MFCD10465684
CAS číslo: 104902-31-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR