Tisk…

2-Bromomethyl-6-methylpyridine 97%

Dodavatel: Apollo Scientific

Synonyms: 2-(Bromomethyl)-6-methylpyridine

OR480737-5G
APOSOR480737-5GEA 2110 CZK
APOSOR480737-5G
2-Bromomethyl-6-methylpyridine 97%
2-Bromomethyl-6-methylpyridine
Vzorec: C₇H₈BrN
MW: 186.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04967229
CAS číslo: 68470-59-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR