Tisk…

6-Nitrobenzo[d]isothiazol-3(2H)-one-1,1-dioxide 95%

Dodavatel: Apollo Scientific

OR480666-25G OR480666-1G OR480666-5G
APOSOR480666-25GEA 3310 CZK
APOSOR480666-25G APOSOR480666-1G APOSOR480666-5G
6-Nitrobenzo[d]isothiazol-3(2H)-one-1,1-dioxide 95%
6-Nitrobenzo[d]isothiazol-3(2H)-one-1,1-dioxide
Vzorec: C₇H₄N₂O₅S
Storage Temperature: Ambient
CAS číslo: 22952-24-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR