Tisk…

Boc-N-methyl-D-alanine 98%

Dodavatel: Apollo Scientific

Synonyms: Boc-N-Me-Ala-OH, N-Boc-N-methyl-L-alanine

OR480585-100G
APOSOR480585-100GEA 3940 CZK
APOSOR480585-100G
Boc-N-methyl-D-alanine 98%
Boc-N-methyl-D-alanine
Vzorec: C₉H₁₇NO₄
MW: 203.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037242
CAS číslo: 16948-16-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR